accessibility menu, dialog, popup

UserName

OHOO

CAS RN 6328-74-1

IUPAC Name: 2-(4-phenoxyphenyl)acetic acid
Synonyms: (4-phenoxyphenyl)acetic acid
Molecular Weight (g/mol): 228.25
Molecular Formula: C14H12O3
InChi Key: VARVNFDGRLLTCI-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=C(OC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results