accessibility menu, dialog, popup

UserName

CAS RN 63214-58-4

ONH2N

CAS RN 63214-58-4

IUPAC Name: 4-(piperidine-1-carbonyl)piperidin-1-ium
Synonyms: 4-(piperidine-1-carbonyl)piperidin-1-ium
Molecular Weight (g/mol): 197.3
Molecular Formula: C11H21N2O
InChi Key: JFHHGGAQABVJIF-UHFFFAOYSA-O
SMILES: O=C(C1CC[NH2+]CC1)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results