accessibility menu, dialog, popup

UserName

HClCH3HNNH2NH2

CAS RN 632-99-5

IUPAC Name: hydrogen 4-[(4-aminophenyl)(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]aniline chloride
Synonyms: fuchsine
Molecular Weight (g/mol): 337.85
Molecular Formula: C20H20ClN3
InChi Key: AXDJCCTWPBKUKL-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=CC(C=CC1=N)=C(C1=CC=C(N)C=C1)C1=CC=C(N)C=C1
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
Color 
  • (7)
  • (1)
Grade 
  • (6)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (9)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (7)
  • (1)
  • (6)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)