accessibility menu, dialog, popup

UserName

CAS RN 632-22-4

CH3NH3CONCH3H3C

CAS RN 632-22-4

IUPAC Name: 1,1,3,3-tetramethylurea
Synonyms: temur tetramethylurea
Molecular Weight (g/mol): 116.16
Molecular Formula: C5H12N2O
InChi Key: AVQQQNCBBIEMEU-UHFFFAOYSA-N
SMILES: CN(C)C(=O)N(C)C
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)