accessibility menu, dialog, popup

UserName

OPHOHH

CAS RN 6303-21-5

IUPAC Name: hydroxy(oxo)phosphanium
Synonyms: hypophosphorous acid
Molecular Weight (g/mol): 64.99
Molecular Formula: H2O2P
InChi Key: GFYXELHUDBJJEV-UHFFFAOYSA-O
SMILES: O[PH+]=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)