accessibility menu, dialog, popup

UserName

NH2SNH2OO

CAS RN 63-74-1

IUPAC Name: 4-aminobenzene-1-sulfonamide
Synonyms: sulfanilamide sulphonamide streptocid sulfanilamide vaginal cream streptocom deseptyl infepan sulphanilamide extra pure tolder P-anilinesulfonamide
Molecular Weight (g/mol): 172.2
Molecular Formula: C6H8N2O2S
InChi Key: FDDDEECHVMSUSB-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)S(N)(=O)=O
Molecular Weight (g/mol) 
  • (9)
  • (10)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
  • (5)
  • (2)
Physical Form 
  • (4)
  • (1)
  • (3)
Certifications/Compliance 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (9)
  • (10)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (5)
  • (2)
  • (4)
  • (1)
  • (3)
  • (3)
  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (1)