accessibility menu, dialog, popup

UserName

H3COONHOOCH3

CAS RN 6290-05-7

IUPAC Name: ethyl 2-[(2-ethoxy-2-oxoethyl)amino]acetate
Synonyms: ethyl 2-[(2-ethoxy-2-oxoethyl)amino]acetate
Molecular Weight (g/mol): 189.21
Molecular Formula: C8H15NO4
InChi Key: LJDNMOCAQVXVKY-UHFFFAOYSA-N
SMILES: CCOC(=O)CNCC(=O)OCC
Molecular Weight (g/mol) 
  • (1)
  • (7)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
Boiling Point 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)