accessibility menu, dialog, popup

UserName

HOOH

CAS RN 629-41-4

IUPAC Name: octane-1,8-diol
Synonyms: 1,8-octanediol
Molecular Weight (g/mol): 146.23
Molecular Formula: C8H18O2
InChi Key: OEIJHBUUFURJLI-UHFFFAOYSA-N
SMILES: OCCCCCCCCO
Quantity 
  • (3)
  • (3)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)