accessibility menu, dialog, popup

UserName

II

CAS RN 629-09-4

IUPAC Name: 1,6-diiodohexane
Synonyms: hexane, 1,6-diiodo-
Molecular Weight (g/mol): 337.97
Molecular Formula: C6H12I2
InChi Key: QLIMAARBRDAYGQ-UHFFFAOYSA-N
SMILES: ICCCCCCI
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)