accessibility menu, dialog, popup

UserName

BrBr

CAS RN 629-03-8

IUPAC Name: 1,6-dibromohexane
Synonyms: hexane, 1,6-dibromo-
Molecular Weight (g/mol): 243.97
Molecular Formula: C6H12Br2
InChi Key: SGRHVVLXEBNBDV-UHFFFAOYSA-N
SMILES: BrCCCCCCBr
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)