accessibility menu, dialog, popup

UserName

CH3NCH3NH2OHSOHOO

CAS RN 6283-63-2

IUPAC Name: N1,N1-diethylbenzene-1,4-diamine; sulfuric acid
Synonyms: N,N-diethyl-1,4-benzenediamine; sulfuric acid
Molecular Weight (g/mol): 262.32
Molecular Formula: C10H18N2O4S
InChi Key: AYLDJQABCMPYEN-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.CCN(CC)C1=CC=C(N)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)