accessibility menu, dialog, popup

UserName

CAS RN 6281-42-1

IUPAC Name: 1-(2-aminoethyl)imidazolidin-2-one
Synonyms: 1-(2-aminoethyl)imidazolidin-2-one
Molecular Weight (g/mol): 129.16
Molecular Formula: C5H11N3O
InChi Key: PODSUMUEKRUDEI-UHFFFAOYSA-N
SMILES: NCCN1CCNC1=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results