accessibility menu, dialog, popup

UserName

H3COCH3

CAS RN 628-81-9

IUPAC Name: 1-ethoxybutane
Synonyms: butyl ethyl ether
Molecular Weight (g/mol): 102.18
Molecular Formula: C6H14O
InChi Key: PZHIWRCQKBBTOW-UHFFFAOYSA-N
SMILES: CCCCOCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results