accessibility menu, dialog, popup

UserName

CAS RN 627-35-0

CH3NHH3C

CAS RN 627-35-0

IUPAC Name: methyl(propyl)amine
Synonyms: N-methylpropylamine
Molecular Weight (g/mol): 73.14
Molecular Formula: C4H11N
InChi Key: GVWISOJSERXQBM-UHFFFAOYSA-N
SMILES: CCCNC
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (2)
  • (1)
  • (3)
  • (3)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)