accessibility menu, dialog, popup

UserName

CAS RN 62637-93-8

H2OH2OCH3NCH3CH3O

CAS RN 62637-93-8

IUPAC Name: N,N-dimethylmethanamine oxide dihydrate
Synonyms: trimethylamine oxide
Molecular Weight (g/mol): 111.14
Molecular Formula: C3H13NO3
InChi Key: PGFPZGKEDZGJQZ-UHFFFAOYSA-N
SMILES: O.O.C[N+](C)(C)[O-]
Percent Purity 
  • (4)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (4)
  • (4)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)