accessibility menu, dialog, popup

UserName

CAS RN 6262-42-6

ClClClCl

CAS RN 6262-42-6

IUPAC Name: tetrachlorocycloprop-1-ene
Synonyms: tetrachlorocycloprop-1-ene
Molecular Weight (g/mol): 177.83
Molecular Formula: C3Cl4
InChi Key: BLZOHTXDDOAASQ-UHFFFAOYSA-N
SMILES: ClC1=C(Cl)C1(Cl)Cl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)