CAS RN 626-76-6
CAS RN 626-76-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
cis-Norbixin, TRC
CAS: 626-76-6 Molecular Formula: C24 H28 O4 Molecular Weight (g/mol): 380.48 Synonym: (2E,4E,6E,8E,10E,12E,14E,16Z,18E)-4,8,13,17-Tetramethyl-2,4,6,8,10,12,14,16,18-eicosanonaenedioic Acid IUPAC Name: (2E,4Z,6E,8E,10E,12E,14E,16E,18E)-4,8,13,17-tetramethylicosa-2,4,6,8,10,12,14,16,18-nonaenedioic acid SMILES: C\C(=C\C=C\C(=C\C=C\C=C(/C)\C=C\C=C(/C)\C=C\C(=O)O)\C)\C=C\C(=O)O