accessibility menu, dialog, popup

UserName

CAS RN 626-67-5

HClCH3N

CAS RN 626-67-5

IUPAC Name: hydrogen 1-methylpiperidine chloride
Synonyms: hydrogen N-methylpiperidine chloride
Molecular Weight (g/mol): 135.64
Molecular Formula: C6H14ClN
InChi Key: QDUXDCXILAPLAG-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN1CCCCC1
Boiling Point 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)