CAS RN 626-56-2
CAS RN 626-56-2
IUPAC Name:
(3S)-3-methylpiperidine
Synonyms:
(3S)-3-methylpiperidine
Molecular Weight (g/mol):
99.18
Molecular Formula:
C6H13N
InChi Key:
JEGMWWXJUXDNJN-LURJTMIESA-N
SMILES:
C[C@H]1CCCNC1
1
–
1
of
1
results
1
3-Methylpiperidine, 97%
CAS: 626-56-2 Molecular Formula: C6H13N Molecular Weight (g/mol): 99.177 MDL Number: MFCD00005994 InChI Key: JEGMWWXJUXDNJN-UHFFFAOYSA-N Synonym: 3-pipecoline,piperidine, 3-methyl,beta-pipecoline,.beta.-pipecoline,3-methyl-piperidine,.beta.-methylpiperidine,beta-methylpiperidine,3-methyl piperidine,3-pipecoline 8ci,3ppc PubChem CID: 79081 IUPAC Name: 3-methylpiperidine SMILES: CC1CCCNC1