accessibility menu, dialog, popup

UserName

CAS RN 62563-07-9

OOBr

CAS RN 62563-07-9

IUPAC Name: 2-(3-bromopropyl)-1,3-dioxolane
Synonyms: 2-(3-bromopropyl)-1,3-dioxolane
Molecular Weight (g/mol): 195.06
Molecular Formula: C6H11BrO2
InChi Key: IQIXIJRPYSOGPY-UHFFFAOYSA-N
SMILES: BrCCCC1OCCO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)