accessibility menu, dialog, popup

UserName

ClI

CAS RN 625-99-0

IUPAC Name: 1-chloro-3-iodobenzene
Synonyms: 3-chloroiodobenzene
Molecular Weight (g/mol): 238.45
Molecular Formula: C6H4ClI
InChi Key: JMLWXCJXOYDXRN-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(I)=C1
Molecular Weight (g/mol) 
  • (9)
  • (3)
Quantity 
  • (1)
  • (3)
  • (4)
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (6)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (9)
  • (3)
  • (1)
  • (3)
  • (4)
  • (3)
  • (1)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)