CAS RN 625-52-5
IUPAC Name:
ethylurea
Synonyms:
ethylurea
Molecular Weight (g/mol):
88.11
Molecular Formula:
C3H8N2O
InChi Key:
RYECOJGRJDOGPP-UHFFFAOYSA-N
SMILES:
CCNC(N)=O
1
–
3
of
3
results
1
Ethylurea, 98%
CAS: 625-52-5 Molecular Formula: C3H8N2O Molecular Weight (g/mol): 88.11 MDL Number: MFCD00007953 InChI Key: RYECOJGRJDOGPP-UHFFFAOYSA-N Synonym: 1-ethylurea,n-ethylurea,urea, ethyl,urea, 1-ethyl,polyurethane,urea, n-ethyl,ethyl urea,pellethane,athylisoharnstoff PubChem CID: 12254 IUPAC Name: ethylurea SMILES: CCNC(N)=O
2
N-Ethylurea, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-Ethylurea, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.