accessibility menu, dialog, popup

UserName

CAS RN 62499-27-8

HO(R)O(S)OOH(R)OH(S)HO(S)HO

CAS RN 62499-27-8

IUPAC Name: (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[4-(hydroxymethyl)phenoxy]oxane-3,4,5-triol
Synonyms: gastrodin
Molecular Weight (g/mol): 286.28
Molecular Formula: C13H18O7
InChi Key: PUQSUZTXKPLAPR-UJPOAAIJSA-N
SMILES: OC[C@H]1O[C@@H](OC2=CC=C(CO)C=C2)[C@H](O)[C@@H](O)[C@@H]1O
Quantity 
  • (2)
  • (2)
  • (2)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results