accessibility menu, dialog, popup

UserName

CAS RN 624-65-7

ClHC

CAS RN 624-65-7

IUPAC Name: 3-chloroprop-1-yne
Synonyms: propargyl chloride
Molecular Weight (g/mol): 74.51
Molecular Formula: C3H3Cl
InChi Key: LJZPPWWHKPGCHS-UHFFFAOYSA-N
SMILES: ClCC#C
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)