CAS RN 623151-90-6
CAS RN 623151-90-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ethiprole Amide, TRC
CAS: 623151-90-6 Molecular Formula: C13H11Cl2F3N4O2S Molecular Weight (g/mol): 415.22 Synonym: 5-Amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-(ethylsulfinyl)-1H-pyrazole-3-carboxamide SMILES: CCS(=O)c1c(N)n(nc1C(=O)N)c2c(Cl)cc(cc2Cl)C(F)(F)F