accessibility menu, dialog, popup

UserName

CAS RN 623-26-7

NN

CAS RN 623-26-7

IUPAC Name: benzene-1,4-dicarbonitrile
Synonyms: 1,4-benzenedicarbonitrile
Molecular Weight (g/mol): 128.13
Molecular Formula: C8H4N2
InChi Key: BHXFKXOIODIUJO-UHFFFAOYSA-N
SMILES: N#CC1=CC=C(C=C1)C#N
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (3)
  • (3)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (6)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)