accessibility menu, dialog, popup

UserName

OHOO

CAS RN 622-04-8

IUPAC Name: 1,3-diphenoxypropan-2-ol
Synonyms: 2-propanol, 1,3-diphenoxy-
Molecular Weight (g/mol): 244.29
Molecular Formula: C15H16O3
InChi Key: NKCVHIPHZCIEFC-UHFFFAOYSA-N
SMILES: OC(COC1=CC=CC=C1)COC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results