CAS RN 62041-46-7
CAS RN 62041-46-7
IUPAC Name:
3-(dimethylamino)-1-(3-methoxyphenyl)prop-2-en-1-one
Synonyms:
3-(dimethylamino)-1-(3-methoxyphenyl)prop-2-en-1-one
Molecular Weight (g/mol):
205.26
Molecular Formula:
C12H15NO2
InChi Key:
QBMMITATXZJWDG-UHFFFAOYSA-N
SMILES:
COC1=CC=CC(=C1)C(=O)C=CN(C)C
1
–
1
of
1
results
1
3-Dimethylamino-1-(3-methoxyphenyl)-2-propen-1-one, 95%, Thermo Scientific™
CAS: 62041-46-7 Molecular Formula: C12H15NO2 Molecular Weight (g/mol): 205.257 MDL Number: MFCD11501639 InChI Key: QBMMITATXZJWDG-UHFFFAOYSA-N Synonym: 2-propen-1-one, 3-dimethylamino-1-3-methoxyphenyl,3-dimethylamino-1-3-methoxyphenyl prop-2-en-1-one,2e-3-dimethylamino-1-3-methoxyphenyl-prop-2-en-1-one,3-dimethylamino-3'-methoxyacrylophenone PubChem CID: 42573796 IUPAC Name: 3-(dimethylamino)-1-(3-methoxyphenyl)prop-2-en-1-one SMILES: CN(C)C=CC(=O)C1=CC(=CC=C1)OC