CAS RN 62020-09-1
IUPAC Name:
methyl 2-methanesulfonylacetate
Synonyms:
methyl 2-methanesulfonylacetate
Molecular Weight (g/mol):
152.16
Molecular Formula:
C4H8O4S
InChi Key:
RQKDASVRZONLNQ-UHFFFAOYSA-N
SMILES:
COC(=O)CS(C)(=O)=O
1
–
1
of
1
results
1
Methyl methylsulfonylacetate, 98+%
CAS: 62020-09-1 Molecular Formula: C4H8O4S Molecular Weight (g/mol): 152.16 MDL Number: MFCD00051842 InChI Key: RQKDASVRZONLNQ-UHFFFAOYSA-N Synonym: methyl methanesulfonylacetate,methyl 2-methanesulfonylacetate,methyl 2-methylsulfonyl acetate,methyl methylsulfonylacetate,methanesulfonylacetic acid methyl ester,acmc-1b4fd,methyl methylsulfonyl acetate,2-methylsulfonyl-1-methoxyethanone,methylsulfonylacetic acid methyl ester,methanesulfonyl acetic acid methyl ester PubChem CID: 2759918 SMILES: COC(=O)CS(C)(=O)=O