accessibility menu, dialog, popup

UserName

NaNaOOOO

CAS RN 62-76-0

IUPAC Name: disodium oxalate
Synonyms: sodium oxalate
Molecular Weight (g/mol): 134
Molecular Formula: C2Na2O4
InChi Key: ZNCPFRVNHGOPAG-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]C(=O)C([O-])=O
Molecular Weight (g/mol) 
  • (8)
  • (31)
Quantity 
  • (2)
  • (1)
  • (4)
  • (9)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (8)
  • (1)
Physical Form 
  • (4)
  • (5)
  • (5)
  • (1)
  • (3)
  • (2)
Grade 
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
Percent Purity 
  • (1)
  • (5)
  • (2)
  • (3)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (3)
  • (4)
  • (3)
  • (1)
  • (3)
  • (7)
  • (9)
  • (3)
  • (2)
  • (8)
  • (31)
  • (2)
  • (1)
  • (4)
  • (9)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (8)
  • (1)
  • (4)
  • (5)
  • (5)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (5)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)