accessibility menu, dialog, popup

UserName

CH3NH2S

CAS RN 62-55-5

IUPAC Name: ethanethioamide
Synonyms: thioacetamide acetic acid thioamide acetic acid, thiono-, amide thioacetimidic acid acetimidic acid, thio- methylthioamide TAA thiacetamid thio- (van) (8CI) acetamide thioactamide
Molecular Weight (g/mol): 75.13
Molecular Formula: C2H5NS
InChi Key: YUKQRDCYNOVPGJ-UHFFFAOYSA-N
SMILES: CC(N)=S
Quantity 
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (5)
  • (3)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (3)
  • (3)
Physical Form 
  • (6)
  • (3)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)
  • (6)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (6)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)
  • (1)
  • (1)