accessibility menu, dialog, popup

UserName

CAS RN 6190-39-2

IUPAC Name: (2R,4R,7R)-N-[(1S,2S,4R,7S)-7-benzyl-2-hydroxy-4-methyl-5,8-dioxo-3-oxa-6,9-diazatricyclo[7.3.0.0²,⁶]dodecan-4-yl]-6-methyl-6,11-diazatetracyclo[7.6.1.0²,⁷.0¹²,¹⁶]hexadeca-1(16),9,12,14-tetraene-4-carboxamide; methanesulfonic acid
Synonyms: dihydroergotamine mesilate 甲磺双氢麦角胺
Molecular Weight (g/mol): 679.79
Molecular Formula: C34H41N5O8S
InChi Key: ADYPXRFPBQGGAH-UMYZUSPBSA-N
SMILES: CS(O)(=O)=O.CN1C[C@@H](C[C@H]2[C@H]1CC1=CNC3=CC=CC2=C13)C(=O)N[C@]1(C)O[C@@]2(O)[C@@H]3CCCN3C(=O)[C@H](CC3=CC=CC=C3)N2C1=O
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (6)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)