CAS RN 619-99-8
CAS RN 619-99-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Ethylhexane, TRC
CAS: 619-99-8 Molecular Formula: C8 H18 Molecular Weight (g/mol): 114.23 Synonym: Hexane, 3-ethyl-,3-Ethylhexane,NSC 74175 IUPAC Name: 3-ethylhexane SMILES: CCCC(CC)CC