accessibility menu, dialog, popup

UserName

HONOO

CAS RN 619-73-8

IUPAC Name: (4-nitrophenyl)methanol
Synonyms: 4-nitrobenzyl alcohol P-nitrobenzyl alcohol paranitrobenzyl alcohol
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: JKTYGPATCNUWKN-UHFFFAOYSA-N
SMILES: OCC1=CC=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)