accessibility menu, dialog, popup

UserName

CAS RN 619-31-8

IUPAC Name: N,N-dimethyl-3-nitroaniline
Synonyms: N,N-dimethyl-3-nitroaniline
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: CJDICMLSLYHRPT-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=CC(=C1)[N+]([O-])=O
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)