accessibility menu, dialog, popup

UserName

ClNN

CAS RN 6188-25-6

IUPAC Name: 6-chloroimidazo[1,2-a]pyridine
Synonyms: 6-chloroimidazo[1,2-a]pyridine
Molecular Weight (g/mol): 152.58
Molecular Formula: C7H5ClN2
InChi Key: XQEGYCZJSVFGEE-UHFFFAOYSA-N
SMILES: ClC1=CN2C=CN=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results