accessibility menu, dialog, popup

UserName

CAS RN 618-48-4

IUPAC Name: 3-chlorobenzamide
Synonyms: 3-chlorobenzamide
Molecular Weight (g/mol): 155.58
Molecular Formula: C7H6ClNO
InChi Key: MJTGQALMWUUPQM-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=CC(Cl)=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)