accessibility menu, dialog, popup

UserName

CAS RN 617-89-0

IUPAC Name: 1-(furan-2-yl)methanamine
Synonyms: furfurylamine
Molecular Weight (g/mol): 97.12
Molecular Formula: C5H7NO
InChi Key: DDRPCXLAQZKBJP-UHFFFAOYSA-N
SMILES: NCC1=CC=CO1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)