accessibility menu, dialog, popup

UserName

CAS RN 617-55-0

H3COOOOCH3OH

CAS RN 617-55-0

IUPAC Name: 1,4-dimethyl 2-hydroxybutanedioate
Synonyms: 1,4-dimethyl 2-hydroxybutanedioate
Molecular Weight (g/mol): 162.14
Molecular Formula: C6H10O5
InChi Key: YSEKNCXYRGKTBJ-UHFFFAOYNA-N
SMILES: COC(=O)CC(O)C(=O)OC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results