accessibility menu, dialog, popup

UserName

CAS RN 617-52-7

H3COOCH2OOCH3

CAS RN 617-52-7

IUPAC Name: 1,4-dimethyl 2-methylidenebutanedioate
Synonyms: 1,4-dimethyl 2-methylidenebutanedioate
Molecular Weight (g/mol): 158.15
Molecular Formula: C7H10O4
InChi Key: ZWWQRMFIZFPUAA-UHFFFAOYSA-N
SMILES: COC(=O)CC(=C)C(=O)OC
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (5)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)