accessibility menu, dialog, popup

UserName

CH3SOOOCH

CAS RN 6165-76-0

IUPAC Name: prop-2-yn-1-yl 4-methylbenzene-1-sulfonate
Synonyms: prop-2-yn-1-yl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 210.25
Molecular Formula: C10H10O3S
InChi Key: LMBVCSFXFFROTA-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OCC#C
Molecular Weight (g/mol) 
  • (1)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (1)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)