accessibility menu, dialog, popup

UserName

HOBOHS

CAS RN 6165-69-1

IUPAC Name: (thiophen-3-yl)boronic acid
Synonyms: thiophen-3-ylboronic acid
Molecular Weight (g/mol): 127.95
Molecular Formula: C4H5BO2S
InChi Key: QNMBSXGYAQZCTN-UHFFFAOYSA-N
SMILES: OB(O)C1=CSC=C1
Percent Purity 
  • (4)
  • (3)
Quantity 
  • (3)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (4)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)