accessibility menu, dialog, popup

UserName

CAS RN 6163-58-2

CH3PH3CCH3

CAS RN 6163-58-2

IUPAC Name: tris(2-methylphenyl)phosphane
Synonyms: tris(2-methylphenyl)phosphane
Molecular Weight (g/mol): 304.37
Molecular Formula: C21H21P
InChi Key: COIOYMYWGDAQPM-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1P(C1=CC=CC=C1C)C1=CC=CC=C1C
Molecular Weight (g/mol) 
  • (3)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)