accessibility menu, dialog, popup

UserName

CAS RN 616-82-0

IUPAC Name: 4-hydroxy-3-nitrobenzoic acid
Synonyms: 4-hydroxy-3-nitrobenzoic acid
Molecular Weight (g/mol): 183.12
Molecular Formula: C7H5NO5
InChi Key: QRYSWXFQLFLJTC-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(O)C(=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results