accessibility menu, dialog, popup

UserName

CAS RN 616-23-9

OHClCl

CAS RN 616-23-9

IUPAC Name: 2,3-dichloropropan-1-ol
Synonyms: 2,3-dichloropropanol
Molecular Weight (g/mol): 128.98
Molecular Formula: C3H6Cl2O
InChi Key: ZXCYIJGIGSDJQQ-UHFFFAOYNA-N
SMILES: OCC(Cl)CCl
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)