accessibility menu, dialog, popup

UserName

H2OH2OH2OH2OMnCH3OOCH3OO

CAS RN 6156-78-1

IUPAC Name: manganese(2+) diacetate tetrahydrate
Synonyms: manganese(2+) diacetate tetrahydrate
Molecular Weight (g/mol): 245.09
Molecular Formula: C4H14MnO8
InChi Key: CESXSDZNZGSWSP-UHFFFAOYSA-L
SMILES: O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O
Physical Form 
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (3)
Grade 
  • (3)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (5)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)