accessibility menu, dialog, popup

UserName

CAS RN 6152-67-6

NaOHSOONH

CAS RN 6152-67-6

IUPAC Name: 4-(phenylamino)benzene-1-sulfonic acid sodium
Synonyms: 4-(phenylamino)benzenesulfonic acid sodium
Molecular Weight (g/mol): 272.27
Molecular Formula: C12H11NNaO3S
InChi Key: VMVSDVIYIMNZBS-UHFFFAOYSA-N
SMILES: [Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (5)
  • (2)
  • (1)
Physical Form 
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
Grade 
  • (3)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (5)
  • (7)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (5)
  • (2)
  • (1)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)