accessibility menu, dialog, popup

UserName

OHOHCl

CAS RN 615-67-8

IUPAC Name: 2-chlorobenzene-1,4-diol
Synonyms: chlorohydroquinone chloroquinol 2-chloro-1,4-hydroxyquinone C6H5ClO2
Molecular Weight (g/mol): 144.55
Molecular Formula: C6H5ClO2
InChi Key: AJPXTSMULZANCB-UHFFFAOYSA-N
SMILES: OC1=CC=C(O)C(Cl)=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (5)
  • (2)