accessibility menu, dialog, popup

UserName

CAS RN 615-60-1

CH3ClH3C

CAS RN 615-60-1

IUPAC Name: 4-chloro-1,2-dimethylbenzene
Synonyms: 3,4-dimethylchlorobenzene
Molecular Weight (g/mol): 140.61
Molecular Formula: C8H9Cl
InChi Key: HNQLMBJUMVLFCF-UHFFFAOYSA-N
SMILES: CC1=CC=C(Cl)C=C1C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)