accessibility menu, dialog, popup

UserName

CAS RN 615-47-4

HHClClNH2NH2H2N

CAS RN 615-47-4

IUPAC Name: dihydrogen benzene-1,2,4-triamine dichloride
Synonyms: dihydrogen 1,2,4-triaminobenzene dichloride
Molecular Weight (g/mol): 196.08
Molecular Formula: C6H11Cl2N3
InChi Key: RKMFFOLUBJFMBQ-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NC1=CC=C(N)C(N)=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)